Learn More
CLC-5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577369
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
Specifications
| CLC-5 | |
| Polyclonal | |
| Unconjugated | |
| Clcn5 | |
| CLCN5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12728, 25749 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P51796, Q9WVD4 | |
| Clcn5 | |
| Peptide CDYENHFNTSKGGEL, corresponding to amino acid residues 401-415 of rat CLC5 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |