Learn More
Emma Safety Footwear Clay (Ruffneck) Safety Shoes
Product Code. 15999071
Click to view available options
Size:
39
40
41
42
43
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15999071
Brand: Emma Safety Footwear 91654639
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reinforced ruff-neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Clay (Ruffneck)
Liner Material Hydro-Tec™ Sanitized Silver
Material Nubuck, Oiled Nubuck
Size 39
Toe Material Steel