Learn More
Claudin-6 Antibody (IM-301) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28245JF585
Product Code. 30173949
  • 431.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Chicken samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-6
Monoclonal
Janelia Fluor 585
claudin 6, claudin-6, skullin
CLDN6
0.1 mL
Stem Cell Markers
9074
Store at 4°C in the dark.
IgG
Flow Cytometry, ELISA, Functional Assay
IM-301
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Chicken
Purified