missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77036C
This item is not returnable.
View return policy
Description
Claudin-1 Polyclonal antibody specifically detects Claudin-1 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Claudin-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| DyLight 650 | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| Antibody was raised against a peptide corresponding to 20 amino acids near the carboxy terminus of human Claudin-1. The immunogen is located within the last 50 amino acids of Claudin-1. Amino Acid Sequence: PRKTTSYPTPRPYPKPAPS | |
| 0.1 mL | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4°C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu