missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin, Light Chain Mouse, Clone: CLC/1421, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1211-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
Clathrin, Light Chain | |
Cytoplasmic | |
CLC/1421 | |
Unconjugated | |
XP_016864515 | |
Mouse | |
31-44kDa | |
Protein A/G Purified | |
RUO | |
1212 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CLTA and CLTB | |
Purified recombinant N-terminal fragment of human Clathrin Light Chain | |
HeLa cells. Placenta or Prostate Carcinoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |