missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Classic Racks for Chest Freezers

Providing safe and secure cold storage. Tenak™ Classic Racks for Chest Freezers can be used with all freezer models.

Brand:  Tenak™ TE21356

Additional Details : Weight : 1.80000kg

 View more versions of this product

Product Code. 15210474

  • 90.85 € / Each
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

  • Racks come with a locking rod to secure cryoboxes
  • Manufactured from stainless steel
  • Sold without cryoboxes
Specifications

Specifications

Freezer Rack
Stainless Steel
1 Pack
140 mm
32 mm
Boxes (14)
Chest freezers
140 x 140 x 504mm
504 mm
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions