Learn More
Tenak™ Classic Rack for Chest Freezers, 130 mm Box Height
Classic Rack is suitable for use with all chest freezer models.
77.80 € - 87.45 €
Specifications
Includes Locking rod
Material Stainless Steel
For Use With (Equipment) Chest Frseezers
Height (Metric) 130 mm (Box)
View More Specs
Products 3
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
15200864
View Documents Promotion Details
Tenak™
TE25102
Boxes (3)
82.35 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-09-2024
to see stock.
Add to basket
15210864
View Documents Promotion Details
Tenak™
TE25104
Boxes (4)
87.45 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-09-2024
to see stock.
Add to basket
15290854
View Documents Promotion Details
Tenak™
TE25100
Boxes (2)
77.80 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Comes with a locking rod to secure the cryoboxes
  • Measure the inside height of you freezer when choosing the right rack
  • Constructed of stainless steel
Specifications
Specifications
Locking rod
Stainless Steel
Chest Frseezers
130 mm (Box)