Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper
Product Code. 16317997 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
200 mL
250 mL
500 mL
1 L
2 L
Certifications/Compliance:
USP Individual
USP Single Batch
Unit Size:
Each
Pack of 2
This item is not returnable. View return policy
Product Code. 16317997
Brand: BRAND™ 236980
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Borosilicate volumetric flask with glass stopper and USP certificate.
  • Borosilicate 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes USP certificate
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Single Batch
Capacity (Metric) 200 mL
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat