Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Product Code. p-7241856 Shop All Heidolph Instruments Products
Click to view available options
Holds:
100 mL Erlenmeyer flasks
1000 mL Erlenmeyer flasks
2000 mL Erlenmeyer flasks
25 mL Erlenmeyer flasks
250 mL Erlenmeyer flasks
50 mL Erlenmeyer flasks
500 mL Erlenmeyer flasks
Unit Size:
Each
Product Code. 10242501
Brand: Heidolph™ 5495100000
Only null left
Add to basket
Add to basket
Clamps for use with various perforated platforms
  • Maximum number of clamps per platform type:
    • 549-51000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-52000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-55000-00
      • For platform 100 - max. 3
      • For platform 1000 - max. 4
      • For platform 2000 - max. 9
    • 549-56000-00
      • For platform 100 - max. 2
      • For platform 1000 - max. 4
      • For platform 2000 - max. 5
    • 549-63000-00
      • For platform 1000 - max. 2
      • For platform 2000 - max. 3
TRUSTED_SUSTAINABILITY
Specifications
Holds 25 mL Erlenmeyer flasks
Product Type Clamp
For Use With (Equipment) Perforated platforms 100, 1000 and 2000
Size 1