missing translation for 'onlineSavingsMsg'
Learn More

Citalopram, Hydrobromide Salt, TRC

Product Code. 17963775
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963775 100 mg 100mg
30137421 50 mg 50mg
17942675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963775 Supplier Toronto Research Chemicals Supplier No. TRCC505000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59729-32-7
Formula Weight 404.09
InChI Formula InChI=1S/C20H21FN2O.BrH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12H,3,10-11,14H2,1-2H3;1H
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
Molecular Formula C20 H21 F N2 O . Br H
Molecular Weight (g/mol) 405.3
Percent Purity >95%
SMILES Br.CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
Synonym Citalopram Hydrobromide, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, hydrobromide (1:1), 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, monohydrobromide (9CI), (±)-Citalopram hydrobromide, Apertia, Celexa, Cheer, Cipram, Cipramil, Citopram, Elopram, Lecital, Lu 10-171B, Lupram, Pramcit, Prisdal, S-Pram, Sepram, Seropram, [3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-isobenzofuran-5-carbonitrile hydrobromide
Chemical Name or Material Citalopram Hydrobromide
Grade HPLC
Show More Show Less

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.