missing translation for 'onlineSavingsMsg'
Learn More

cis-6-Nonenal, 94%, Thermo Scientific Chemicals

Product Code. 11368065
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11368065

Brand: Thermo Scientific Alfa Aesar L08260.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a flavor component of muskmelon fruit.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Air sensitive. Store away air and from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2277-19-2
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00014683
InChI Key RTNPCOBSXBGDMO-ARJAWSKDSA-N
Synonym cis-6-nonenal, z-6-nonenal, z-non-6-enal, cis-6-nonen-1-al, 6-nonenal, z, 6-nonenal, cis, 6z-non-6-enal, 6-cis-nonenal, 6-nonenal, 6z, fema no. 3580
PubChem CID 5362720
IUPAC Name (Z)-non-6-enal
SMILES CCC=CCCCCC=O

Specifications

Density 0.847
Boiling Point 72°C (10 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.445
Quantity 2 g
Beilstein 2323664
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 140.22
Percent Purity 94%
Chemical Name or Material cis-6-Nonenal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.