missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%

Product Code. p-7027893
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11461487

Brand: Thermo Scientific Alfa Aesar B24951.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129-64-6 | C9H8O3 | 164.16 g/mol

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-64-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
InChI Key KNDQHSIWLOJIGP-UHFFFAOYNA-N
Synonym cis-5-norbornene-endo-2,3-dicarboxylic anhydride
PubChem CID 126963424
IUPAC Name 4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1OC(=O)C2C3CC(C=C3)C12

Specifications

Melting Point 164°C to 167°C
Density 1.417
Quantity 5 g
Beilstein 83657
Sensitivity Moisture sensitive
Merck Index 14,1796
Solubility Information It decomposes in water.
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.