Learn More
cis-4-Decenal, TRC
Product Code. 30359758
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359758 50 mg 50mg
30360187 500 mg 500mg
30355965 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30359758 Supplier Toronto Research Chemicals Supplier No. TRCC47583050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21662-09-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.24
IUPAC Name (Z)-dec-4-enal
SMILES CCCCC\C=C/CCC=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H18O/c1-2-3-4-5-6-7-8-9-10-11/h6-7,10H,2-5,8-9H2,1H3/b7-6-
Formula Weight 154.1358
Chemical Name or Material cis-4-Decenal