Learn More
cis-3-Dodecen-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | cis-3-Dodecen-1-ol |
| CAS | 32451-95-9 |
| Molecular Formula | C12H24O |
| Quantity | 1 g |
| Synonym | (3Z)-3-Dodecen-1-ol, (Z)-3-Dodecen-1-ol |
| InChI Formula | InChI=1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h9-10,13H,2-8,11-12H2,1H3/b10-9- |
| SMILES | CCCCCCCC\C=C/CCO |
| IUPAC Name | (Z)-dodec-3-en-1-ol |
| Molecular Weight (g/mol) | 184.32 |
| Formula Weight | 184.1827 |