Learn More
cis-1-Chloro-1,5-hexadiene, TRC
Specifications
Specifications
| Chemical Name or Material | cis-1-Chloro-1,5-hexadiene |
| CAS | 27934-74-3 |
| Molecular Formula | C6H9Cl |
| Quantity | 2.5 mg |
| Synonym | (Z)-1-Chloro-1,5-hexadiene |
| InChI Formula | InChI=1S/C6H9Cl/c1-2-3-4-5-6-7/h2,5-6H,1,3-4H2/b6-5- |
| SMILES | C=CCC/C=C\Cl |
| IUPAC Name | (1Z)-1-chlorohexa-1,5-diene |
| Molecular Weight (g/mol) | 116.59 |
| Formula Weight | 116.0393 |