Learn More
cis-1-Chloro-1,5-hexadiene, TRC
Product Code. 30350433
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350433 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30350433 Supplier Toronto Research Chemicals Supplier No. TRCC36401525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material cis-1-Chloro-1,5-hexadiene
CAS 27934-74-3
Molecular Formula C6H9Cl
Quantity 25 mg
Synonym (Z)-1-Chloro-1,5-hexadiene
InChI Formula InChI=1S/C6H9Cl/c1-2-3-4-5-6-7/h2,5-6H,1,3-4H2/b6-5-
SMILES C=CCC/C=C\Cl
IUPAC Name (1Z)-1-chlorohexa-1,5-diene
Molecular Weight (g/mol) 116.59
Formula Weight 116.0393