Learn More
cis-1,2-Cyclohexanedicarboxylic acid, 98%
Product Code. 11348445
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348445 25 g 25g
11358445 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11348445 Supplier Thermo Scientific Alfa Aesar Supplier No. L10466.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 610-09-3 | C8H10O4 | 170.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610-09-3
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00064949
InChI Key QSAWQNUELGIYBC-OLQVQODUSA-L
Synonym cis-1,2-cyclohexanedicarboxylic acid, cis-cyclohexane-1,2-dicarboxylic acid, cis-hexahydrophthalic acid, 1r,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2r-cyclohexane-1,2-dicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2s-rel, cis-1,2-cyclohexanedicarboxylicacid, ksc497o9d, cis-+/--perhydrophthalic acid, cis-cyclohexane-1,2-dicarboxylicacid
PubChem CID 245564
IUPAC Name (1S,2R)-cyclohexane-1,2-dicarboxylic acid
SMILES [O-]C(=O)[C@H]1CCCC[C@H]1C([O-])=O
Specifications
Melting Point 189°C to 192°C
Quantity 25 g
Beilstein 2049754
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic acid