missing translation for 'onlineSavingsMsg'
Learn More

Cinnamic Acid-13C3, TRC

Product Code. 17993145
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993145 50 mg 50mg
17950965 1 mg 1mg
17911645 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993145 Supplier Toronto Research Chemicals Supplier No. TRCC44203250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C613C3H8O2
Molecular Weight (g/mol) 151.14
Synonym 3-Phenyl-2-propenoic Acid-1,2,3-13C3, 3-Phenylacrylic Acid-1,2,3-13C3, NSC 623441-1,2,3-13C3, NSC 9189-1,2,3-13C3, Phenylacrylic Acid-1,2,3-13C3, β-Phenylacrylic Acid-1,2,3-13C3;
IUPAC Name (E)-3-phenyl(1,2,3-13C3)prop-2-enoic acid
SMILES O[13C](/[13C]=[13C]/C1=CC=CC=C1)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+/i6+1,7+1,9+1
Chemical Name or Material Cinnamic Acid-13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.