Learn More
Cimifugin, MedChemExpress
Product Code. 16459048
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459048 10 mg 10mg
16431468 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16459048 Supplier Medchem Express Supplier No. HYN0634
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.
Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.
Specifications
Chemical Name or Material Cimifugin
CAS 37921-38-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18O6
Quantity 10 mg
Synonym Cimitin
Solubility Information DMSO : 100 mg/mL (326.47 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (108.81 mM; Need ultrasonic)
SMILES O=C1C=C(CO)OC2=CC(O[C@H](C(C)(O)C)C3)=C3C(OC)=C12
Molecular Weight (g/mol) 306.31
Formula Weight 306.31
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation