missing translation for 'onlineSavingsMsg'
Learn More

Cimifugin, MedChemExpress

Product Code. 16431468
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431468 5 mg 5mg
16459048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431468 Supplier Medchem Express Supplier No. HYN0634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.

Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.

Spécification

Chemical Name or Material Cimifugin
CAS 37921-38-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18O6
Quantity 5 mg
Synonym Cimitin
Solubility Information DMSO : 100 mg/mL (326.47 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (108.81 mM; Need ultrasonic)
SMILES O=C1C=C(CO)OC2=CC(O[C@H](C(C)(O)C)C3)=C3C(OC)=C12
Molecular Weight (g/mol) 306.31
Formula Weight 306.31
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.