Learn More
Cilostazol, MedChemExpress
Product Code. 16463377
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16463377
Brand: Medchem Express HY17464
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Cilostazol (OPC 13013) is a potent and selective inhibitor of phosphodiesterase (PDE) 3A, the isoform of PDE 3 in the cardiovascular system, with an IC50 of 0.2 μM.
Cilostazol (OPC 13013) is a potent and selective inhibitor of phosphodiesterase (PDE) 3A, the isoform of PDE 3 in the cardiovascular system, with an IC50 of 0.2 μM.
Specifications
Chemical Name or Material Cilostazol
CAS 73963-72-1
Color White
Purity Grade Notes Research
Molecular Formula C20H27N5O2
Quantity 50 mg
Synonym OPC 13013
Solubility Information DMSO : 50 mg/mL (135.33 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=C(OCCCCC3=NN=NN3C4CCCCC4)C=C2)CC1
Molecular Weight (g/mol) 369.46
Formula Weight 369.46
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation