Learn More
Cilastatin-15N-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Cilastatin-15N-d3 |
| CAS | 2738376-83-3 |
| Molecular Formula | C16 D3 H23 15N N O5 S |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C16H26N2O5S/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23)/b12-6-/t10-,11+/m1/s1/i9D2,11D,17+1 |
| SMILES | [2H]C([2H])(SCCCC\C=C(/NC(=O)[C@H]1CC1(C)C)\C(=O)O)[C@]([2H])([15NH2])C(=O)O |
| IUPAC Name | (Z)-7-[(2R)-2-(15N)azanyl-2-carboxy-1,1,2-trideuterioethyl]sulfanyl-2-[[(1S)-2,2-dimethylcyclopropanecarbonyl]amino]hept-2-enoic acid |
| Molecular Weight (g/mol) | 362.465 |
| Formula Weight | 362.172 |