missing translation for 'onlineSavingsMsg'
Learn More

CID 2011756, MedChemExpress

Product Code. 16435698
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435698

Brand: Medchem Express HY13454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CID 2011756 is an ATP competitive PKD inhibitor, with an IC50 of 3.2 μM for PKD1 in cell free assay, and also shows cellular pan-PKD inhibitory activity against PKD2 and PKD3 (IC50, 0.6 and 0.7 μM, respectively). CID 2011756 also has antitumor activity.

CID 2011756 is an ATP competitive PKD inhibitor, with an IC50 of 3.2 μM for PKD1 in cell free assay, and also shows cellular pan-PKD inhibitory activity against PKD2 and PKD3 (IC50, 0.6 and 0.7 μM, respectively). CID 2011756 also has antitumor activity.

Specifications

Chemical Name or Material CID 2011756
CAS 638156-11-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H21ClN2O3
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (50.39 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C2=CC=CC(Cl)=C2)O1)NC3=CC=C(CN4CCOCC4)C=C3
Molecular Weight (g/mol) 396.87
Formula Weight 396.87
Percent Purity 96.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.