missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CIC Polyclonal Antibody

Product Code. 13278518
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13278518 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13278518 Supplier Invitrogen™ Supplier No. PA524198

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CIC Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CIC
Gene Accession No. Q924A2, Q96RK0
Gene Alias 1200010B10Rik; Capicua; capicua homolog; capicua homolog (Drosophila); capicua transcriptional repressor; CIC; KIAA0306; mKIAA0306; Protein capicua homolog; protein capicua homolog; LOW QUALITY PROTEIN: protein capicua homolog
Gene Symbols CIC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 174-203 amino acids from the N-terminal region of human CIC.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23152, 71722
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.