missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST6 Monoclonal Antibody (OTI1D1)

Description
CHST6 Monoclonal Antibody for Western Blot
Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the transfer of sulfate to position 6 of non-reducing N-acetylglucosamine (GlcNAc) residues of keratan. Mediates sulfation of keratan in cornea. Keratan sulfate plays a central role in maintaining corneal transparency. Acts on the non-reducing terminal GlcNAc of short and long carbohydrate substrates that have poly-N- acetyllactosamine structures.
Specifications
Specifications
| Antigen | CHST6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CHST6 |
| Gene Accession No. | Q9GZX3 |
| Gene Alias | carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6; carbohydrate sulfotransferase 5; carbohydrate sulfotransferase 6; carbohydrate sulfotransferase 6; C-GlcNAc6ST; CHST6; corneal N-acetylglucosamine 6-sulfotransferase; corneal N-acetylglucosamine-6-O-sulfotransferase; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta; glcNAc6ST-5; gn6st-5; GST4-beta; hCGn6ST; LOC489707; MCDC1; N-acetylglucosamine 6-O-sulfotransferase 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?