Learn More
Proteintech ChromoTek pSpot4 Vector
Product Code. 17393323
Click to view available options
Quantity:
1.25 μg
Unit Size:
1.25µg
Product Code. 17393323
Brand: Proteintech ev4
Please to purchase this item. Need a web account? Register with us today!
pSpot4 vector, E.coli, Spot N-term, Amp., low expression.
  • The plasmid pSpot4 is an expression vector for Spot-Tag™ fusion proteins in E. coli. After cloning the protein of interest (POI) into the multiple cloning site (MCS) provided by pSpot4, the Spot-Tag™ (sequence: PDRVRAVSHWSS) will be fused to the N-terminus of the POI.
Spezifikation
Quantity 1.25 μg