missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Booster ATTO647N

Product Code. p-7235992
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
This item is not returnable. View return policy

Product Code. 17343343

Brand: Proteintech GBA647N10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GFP VHH conjugated to ATTO 647N.

  • anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy

Specifications

Clonality Monoclonal
Quantity 10 μL
For Use With (Application) Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
Conjugate ATTO647N
Target Specificity AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP
Content And Storage Shipped at ambient temperature. Upon receipt store at +4™C. Stable for 6 month. Do not freeze. Protect from light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.