Learn More
Lenz Laborglasintrumente Chromatographic Columns with Frit and NS Socket with PTFE or Needle-Valve Stopcock
Click to view available options
Diameter (Metric) Outer:
10 mm
15 mm
20 mm
30 mm
Unit Size:
Each
Beskrivelse
- Chromatographic columns
- With sintered frit (porosity: 0)
- With PTFE stopcock or needle-valve stopcock (bore: 0 to 2.5 mm)
- Made of DURAN™ tubing
Tekniske data
Tekniske data
| Capacity (Metric) Reservoir | 125 mm |
| Description | Needle-valve stopcock |
| Length (Metric) | 400 mm |
| Diameter (Metric) Outer | 20 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion