missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Chromate Test Kits
Click to view available options
Range:
0.05 to 2 mg/L
Unit Size:
Pack of 25
Description
- For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
- For waste water, drinking water
- Chromate (chromium VI and total chromium) cell test kit
- Photometric test method
- This test is highly accurate and easy to use
Specifications
Specifications
| Analytes | Diphenylcarbazide |
| For Use With (Application) | For waste water, drinking water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Range | 0.05 to 2 mg/L |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | Chromate (chromium VI and total chromium) cell test kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction