Learn More
Chromane, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydro-2H-1-benzopyran |
| CAS | 493-08-3 |
| Molecular Formula | C9H10O |
| Quantity | 50 mg |
| Synonym | 3,4-Dihydro-2H-1-benzopyran, Chromane |
| InChI Formula | InChI=1S/C9H10O/c1-2-6-9-8(4-1)5-3-7-10-9/h1-2,4,6H,3,5,7H2 |
| SMILES | C1COc2ccccc2C1 |
| IUPAC Name | 3,4-dihydro-2H-chromene |
| Molecular Weight (g/mol) | 134.17 |
| Formula Weight | 134.0732 |