Learn More
Chroman 1, MedChemExpress
Product Code. 16479067
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16479067
Brand: Medchem Express HY15392
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Chroman 1 is a highly potent and selective ROCK inhibitor. Chroman 1 is more potent against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM). Chroman 1 also has inhibitory activity against MRCK, with an IC50 of 150 nM.
Chroman 1 is a highly potent and selective ROCK inhibitor. Chroman 1 is more potent against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM). Chroman 1 also has inhibitory activity against MRCK, with an IC50 of 150 nM.
Specifications
Chemical Name or Material Chroman 1
CAS 1273579-40-0
Color Pink
Purity Grade Notes Research
Molecular Formula C24H28N4O4
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (114.55 mM)
SMILES O=C([C@@H]1COC2=CC=C(OC)C=C2C1)NC3=CC=C(C4=CNN=C4)C=C3OCCN(C)C
Molecular Weight (g/mol) 436.5
Formula Weight 436.5
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus