Learn More
Chromalite™ A860/5952 Macroporous Polyacrylic Resin Crosslinked with Divinylbenzene Strong Anion Exchanger, Purolite™
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
Description
Ideal for isolation of proteins, especially for proteins from milk and dairy products.
- Optimal for lactoferrin purification from whey
- Hydrophilic, protein-compatible matrix
- High rigidity
- Suitable for use in industrial columns
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
| Physical Form | Spherical Beads |
| Particle Size | 300 to 850 μm |
| Volume Total Capacity | 0.8 eq/L |
| Quantity | 1 L |
| Product Line | A860/5952 Macroporous Polyacrylic Resin |
| Functionality/Interaction Type | Quaternary Ammonium |
| pH Range | 1 to 14 |