Learn More
Macherey-Nagel™ CHROMABOND™ Complete Vacuum Manifold
Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
up to 12 Columns
up to 16 LV Columns
up to 24 Columns
Unit Size:
Each
Description
The complete CHROMABOND Vacuum Manifold consists of:
- Rectangular glass cabinet with polypropylene lid and lid gasket (2 sizes available):
- Small holds up to 12 CHROMABOND™ columns or CHROMAFIX™ cartridges
- Large holds up to 16 CHROMABOND™ LV columns or up to 24 CHROMABOND™ columns or CHROMAFIX™ cartridges
- Removable needles on lower side of the lid
- Vacuum gauge for reading the vacuum pressure in the chamber
- Control valve for adjustment of the vacuum in the chamber
- Replaceable valves with caps that allow individual vacuum control of each SPE column
- Variable rack with exchangeable partitions, which accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials, etc.
Specifications
Specifications
| For Use With (Application) | Solid Phase Extraction |
| For Use With (Equipment) | up to 16 LV Columns |
| Type | Vacuum Manifold |
| Quantity | 1 Each |