Learn More
Macherey-Nagel™ Chromabond™ NH2/C18 Polypropylene SPE Column
Product Code. 10368261 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
500 mg/1 g
500/500 mg
Unit Size:
Pack of 30
Product Code. 10368261
Brand: Macherey-Nagel™ 730620
Only null left
Add to basket
Add to basket
Specifications
Column Capacity (Metric) 6 mL
Column Type SPE
Bed Weight 500 mg/1 g
For Use With (Application) Solid Phase Extraction Analysis of PAH
Sorbent Chromabond NH2/C18
Type Solid Phase Extraction Column