Learn More
Macherey-Nagel™ Chromabond™ C6 H5 Polypropylene SPE Column
Product Code. 10786332 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 10786332
Brand: Macherey-Nagel™ 730083
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Base Material Silica
Carbon Load 8%
Column Capacity (Metric) 1 mL
Column Type SPE
Bed Weight 100 mg
Particle Size 45 μm
Type Solid Phase Extraction Column
Pore Size 60 μm
Surface Area (Metric) 500 m2/g