missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHR 2797
GREENER_CHOICE

Product Code. 18015812
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 18015812

Brand: Tocris Bioscience™ 3595/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aminopeptidase inhibitor

CHR 2797 is an aminopeptidase inhibitor (IC50 values are 100, 150, 220, > 1000, > 5000, > 10000 and > 30000 nM for LAP, PuSA, aminopeptidase N, aminopeptidase B, PILSAP, LTA4 hydrolase and MetAP2, respectively). Potently inhibits tumor cell proliferation in a variety of tumor cell lines in vitro and in vivo. Also inhibits neprilysin (IC50 = 900 nM). Exhibits antinociceptive activity and enhances the efficacy of Morphine (Cat. No. 5158) in the hot-plate and tail-flick assays, via an action on the peripheral nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type CHR 2797
Purity >98%
Molecular Weight (g/mol) 406.48
Inhibitors CHR 2797

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.