Learn More
Choline-d9 Bromide (N,N,N-trimethyl-d9), CDN
Specifications
Specifications
| Chemical Name or Material | Choline-D9 (N,N,N-trimethyl-D9) Bromide |
| CAS | 285979-71-7 |
| Molecular Formula | C5 D9 H5 N O . Br |
| Quantity | 0.25 g |
| Synonym | Ethanaminium, 2-hydroxy-N,N,N-tri(methyl-d3)-, bromide (9 CI), Choline-tri(methyl-d3) bromide, Methyl-D9-choline bromide, Choline-d9 Bromide, Choline-D9 (N,N,N-trimethyl-D9) Bromide, 2-Hydroxy-N,N,N-tri(methyl-d3)ethanaminium bromide (1:1) |
| InChI Formula | InChI=1 S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+](CCO)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-hydroxyethyl-tris(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 193.1302 |
| Formula Weight | 192.0824 g/mol |
| Show More |