Learn More
Choline-d4 Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Choline-d4 Chloride |
| CAS | 285979-70-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 D4 H10 N O . Cl |
| Quantity | 50 mg |
| Synonym | Choline-1,1,2,2-d4 Chloride, Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-trimethyl-chloride, Choline chloride-1,1,2,2-d4, Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-trimethyl-, chloride (9CI), 2-Hydroxy-N,N,N-trimethylethan-1,1,2,2-d4-aminium chloride |
| InChI Formula | InChI=1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i4D2,5D2; |
| SMILES | [Cl-].[2H]C([2H])(O)C([2H])([2H])[N+](C)(C)C |
| IUPAC Name | trimethyl-(1,1,2,2-tetradeuterio-2-hydroxyethyl)azanium;chloride |
| Molecular Weight (g/mol) | 143.6484 |
| Show More |
Product Title
Ser du en mulighed for forbedring?