Learn More
Cholestyramine (Technical Grade), TRC
Product Code. 17918214
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17918214 10 g 10g
17939734 5 g 5g
17968964 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17918214 Supplier Toronto Research Chemicals Supplier No. TRCC43255510G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 11041-12-6
Molecular Formula (C26H40ClN)n
Molecular Weight (g/mol) 435.1
Synonym Cholestyramine Chloride, Cholestyramine Hydroxide, Cholestyramine Resin, Cholybar, Colestyramin, Colestyramine, Cuemid, LoCholest, MK 135, Quantalan, Questran, Questran Light;
IUPAC Name 1-(4-(5-(3-(sec-butyl)phenyl)hexan-3-yl)phenyl)-N,N,N-trimethylmethanaminium chloride
SMILES CC(CC(CC)C1=CC=C(C[N+](C)(C)C)C=C1)C2=CC=CC=C2.CC(CC)C.[Cl-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C22H32N.C5H12.ClH/c1-6-20(16-18(2)21-10-8-7-9-11-21)22-14-12-19(13-15-22)17-23(3,4)5;1-4-5(2)3;/h7-15,18,20H,6,16-17H2,1-5H3;5H,4H2,1-3H3;1H/q+1;;/p-1
Chemical Name or Material Colestyramine