Learn More
Chlorpyrifos-d10 (diethyl-d10), CDN
Specifications
Specifications
| Chemical Name or Material | Chlorpyrifos D10 |
| CAS | 285138-81-0 |
| Molecular Formula | C9 2H10 H Cl3 N O3 P S |
| Quantity | 0.005 g |
| Synonym | Phosphorothioic acid, O,O-di(ethyl-d5) O-(3,5,6-trichloro-2-pyridinyl) ester (9 CI), Chlorpyrifos D10, Chlorpyrifos (diethyl-D10) |
| InChI Formula | InChI=1 S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OP(=S)(Oc1nc(Cl)c(Cl)cc1Cl)OC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 360.65 |
| Formula Weight | 358.9891 g/mol |
| Show More |