Learn More
Chlorprothixene-d6, TRC
Product Code. 30359987
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359987 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359987 Supplier Toronto Research Chemicals Supplier No. TRCC4249125MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorprothixene-d6
CAS 1246912-83-3
Molecular Formula C18H12D6ClNS
Quantity 5 mg
Synonym (3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine-d6, (Z)-2-Chloro-N,N-dimethyl-thioxanthene-delta9,gamma-propylamine-d6, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine-d6, CPT-d6, 3-(2-Chlorothioxanthen-9-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine
InChI Formula InChI=1S/C18H18ClNS/c1-20(2)11-5-7-14-15-6-3-4-8-17(15)21-18-10-9-13(19)12-16(14)18/h3-4,6-10,12H,5,11H2,1-2H3/b14-7-/i1D3,2D3
SMILES [2H]C([2H])([2H])N(CC\C=C/1\c2ccccc2Sc3ccc(Cl)cc13)C([2H])([2H])[2H]
IUPAC Name (3Z)-3-(2-chlorothioxanthen-9-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine
Molecular Weight (g/mol) 321.9