Learn More
Chloromethyl Propyl Carbonate, TRC
Product Code. 30366496
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366496 5 g -
30360412 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30366496 Supplier Toronto Research Chemicals Supplier No. TRCC3692255G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35273-90-6
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.58
Synonym Chloromethyl Carbonic Acid Propyl Ester
IUPAC Name chloromethyl propyl carbonate
SMILES CCCOC(=O)OCCl
Specifications
Quantity 5 g
InChI Formula InChI=1S/C5H9ClO3/c1-2-3-8-5(7)9-4-6/h2-4H2,1H3
Formula Weight 152.024
Chemical Name or Material Chloromethyl Propyl Carbonate