missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)cyclopropane, 97%

Product Code. p-7044533
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11357076

Brand: Thermo Scientific Alfa Aesar L06986.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5911-08-0 | C4H7Cl | 90.55 g/mol

(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
SMILES ClCC1CC1

Specifications

Melting Point -91°C
Density 0.98
Boiling Point 87°C to 89°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.436
Quantity 5 g
UN Number UN1993
Beilstein 635697
Solubility Information Not miscible in water.
IUPAC Name chloromethylcyclopropane
Formula Weight 90.55
Percent Purity 97%
Chemical Name or Material (Chloromethyl)cyclopropane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.