Learn More
β-Chloro-L-alanine, MedChemExpress
Product Code. 16475086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16475086 Supplier Medchem Express Supplier No. HY107373
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress β-Chloro-L-alanine is a bacteriostatic amino acid analog which inhibits a number of enzymes, including threonine deaminase and alanine racemase.
β-Chloro-L-alanine is a bacteriostatic amino acid analog which inhibits a number of enzymes, including threonine deaminase and alanine racemase.
Specifications
Chemical Name or Material β-Chloro-L-alanine
Physical Form Solid
CAS 2731-73-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H6ClNO2
For Use With (Application) COVID-19-immunoregulation
Synonym L-β-Chloroalanine
Solubility Information H2O : ≥ 30 mg/mL (242.84 mM)
SMILES OC([C@@H](N)CCl)=O
Molecular Weight (g/mol) 123.54
Formula Weight 123.54
Percent Purity 97.0%
Grade Research