Learn More
Chlormadinol Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Chlormadinol Acetate |
| CAS | 3114-44-1 |
| Molecular Formula | C23 H31 Cl O4 |
| Quantity | 5 mg |
| Synonym | Pregna-4,6-dien-20-one, 17-(acetyloxy)-6-chloro-3-hydroxy-, (3β)-, Pregna-4,6-dien-20-one, 6-chloro-3β,17-dihydroxy-, 17-acetate (7CI,8CI), (3β)-17-(Acetyloxy)-6-chloro-3-hydroxypregna-4,6-dien-20-one, 17α-Acetoxy-6-chloro-3β-hydroxypregna-4,6-dien-20-one, 3β-Hydroxychlormadinone acetate, 6-Chloro-3β,17-dihydroxypregna-4,6-dien-20-one 17-acetate, AY 11497, Chlormadinol acetate, 17α-Acetoxy-6-chloro-3.beta.-hydroxypregna-4,6-dien-20-one |
| InChI Formula | InChI=1S/C23H31ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,15-18,27H,5-10H2,1-4H3/t15-,16+,17-,18-,21+,22-,23-/m0/s1 |
| SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(3S,8R,9S,10R,13S,14S,17R)-17-acetyl-6-chloro-3-hydroxy-10,13-dimethyl-1,2,3,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Molecular Weight (g/mol) | 406.94 |
| Formula Weight | 406.1911 |