Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 250420
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10489243
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
DIN 38408 G4, EN ISO 7393
Disinfectants, wastewater, drinking water
Free and total Chlorine
0.03 to 6.00 Cl2, free and total
Cell Test Kit
5 mL
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter
150 Pack
DPD