Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 250420
Additional Details : Weight : 0.25000kg
Specifications
DIN 38408 G4, EN ISO 7393 | |
Disinfectants, wastewater, drinking water | |
Free and total Chlorine | |
0.03 to 6.00 Cl2, free and total | |
Cell Test Kit |
5 mL | |
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter | |
150 Pack | |
DPD |