Learn More
Chlorguanide-d4 Hydrochloride, TRC
Product Code. 30355621
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355621 1 mg -
30361510 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355621 Supplier Toronto Research Chemicals Supplier No. TRCC3295021MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1189671-34-8
Molecular Formula C11H13D4Cl2N5
Molecular Weight (g/mol) 294.22
Synonym N-(4-Chlorophenyl-d4)-N'-(1-methylethyl)-imidodicarbonimidic Diamide Hydrochloride, 1-(p-Chlorophenyl-d4)-5-isopropylbiguanide Hydrochloride, Bigumal-d4, Chlorguanid-d4, Chloroguanide-d4, Paludrin-d4, Proguanil-d4
IUPAC Name 1-((l2-azaneylidene)((4-chlorophenyl-2,3,5,6-d4)amino)methyl)-3-isopropylguanidine
SMILES CC(C)NC(NC(NC1=C([2H])C([2H])=C(Cl)C([2H])=C1[2H])=N)=N.Cl
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H/i3D,4D,5D,6D;
Formula Weight 293.11
Chemical Name or Material Chlorguanide-d4 Hydrochloride