Learn More
Chitinase-IN-2, MedChemExpress
Description
Chitinase-IN-2 is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide
Specifications
Specifications
| Chemical Name or Material | Chitinase-IN-2 |
| Physical Form | Oil |
| CAS | 1579991-63-1 |
| Color | Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H21N5O2S |
| Solubility Information | DMSO : ≥ 53 mg/mL (134.01 mM) |
| SMILES | O=C(C1=CC=CC2=C1C3=CC=C2N(C)C)N(CCNCC4=NN=C(C)S4)C3=O |
| Molecular Weight (g/mol) | 395.48 |
| Show More |