missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicoric acid, MedChemExpress
Description
Chicoric acid inhibits cell viability and induces mitochondria-dependent apoptosis in 3T3-L1 preadipocytes through ROS-mediated PI3K/Akt and MAPK signaling pathways. Chicoric acid increases glucose uptake, improves insulin resistance, and attenuates glucosamine-induced inflammation. Chicoric acid has antidiabetic properties and antioxidant, anti-inflammatory effects.
Specifications
Specifications
| Chemical Name or Material | Chicoric acid |
| CAS | 6537-80-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18O12 |
| Quantity | 25 mg |
| Synonym | Cichoric acid Dicaffeoyltartaric acid |
| Solubility Information | H2O : 100 mg/mL (210.81 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.11 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@H](OC(/C=C/C1=CC=C(O)C(O)=C1)=O)[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C(O)=O |
| Molecular Weight (g/mol) | 474.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?