Learn More
Tocris Bioscience™ CHDI 113
Description
CHDI 113 is a potent huntingtin (HTT) pre-mRNA splicing modulator (EC50 = 66 nM). It lowers HTT protein in both human Huntington's disease (HD) stem cells and in the brain of BACHD (bacterial artificial chromosome-mediated transgenic) mouse model. CHDI 113 is brain penetrant and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-[(3R)-3-(Cyclopropylamino)-1-pyrrolidinyl]-N-(8-fluoro-2-methylimidazo[1,2-a]pyridin-6-yl)-2-pyrazinecarboxamide |
| CAS | 2743336-70-9 |
| Quantity | 5 mg |
| Target | Huntingtin |
| Molecular Formula | C20H22FN7O |
| Purity | 0.98 |